siRNA Details
Gene Symbol: LOC101929541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125983.1 | 3 | 403 | |||
NR_125984.1 | 4 | 282 | ||||
GenBank mRNA | BX099738.1 | - | 4 | 282 | ||
F26527.1 | - | 3 | 213 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000412221.5 | - | 2 | 215 | Chr. 1 - 212225280 - 212226538 [-] on Build GRCh38 |
