siRNA Details
| Gene Symbol: BAZ2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110586.1 | 3 | 2492 | |||
| GenBank mRNA | BX537650.1 | - | 3 | 2492 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000607836.1 | - | 3 | 2492 | Chr. 2 - 159693201 - 159712435 [-] on Build GRCh38 | |

BAZ2B
Genomic Map