siRNA Details
| Gene Symbol: HAGLROS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110457.1 | 2 | 506 | |||
| GenBank mRNA | AA430081.1 | - | 2 | 432 | ||
| HY007937.1 | - | 2 | 506 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HAGLROS| Gene Symbol: HAGLROS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110457.1 | 2 | 506 | |||
| GenBank mRNA | AA430081.1 | - | 2 | 432 | ||
| HY007937.1 | - | 2 | 506 | |||