siRNA Details
| Gene Symbol: LOC101929378 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110249.2 | 4 | 790 | |||
| NR_110250.2 | 4 | 840 | ||||
| GenBank mRNA | BC032407.1 | - | 4 | 779 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929378| Gene Symbol: LOC101929378 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110249.2 | 4 | 790 | |||
| NR_110250.2 | 4 | 840 | ||||
| GenBank mRNA | BC032407.1 | - | 4 | 779 | ||