siRNA Details
| Gene Symbol: LOC101927640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121193.1 | 3 | 402 | |||
| GenBank mRNA | AI148060.1 | - | 3 | 402 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927640| Gene Symbol: LOC101927640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121193.1 | 3 | 402 | |||
| GenBank mRNA | AI148060.1 | - | 3 | 402 | ||