siRNA Details
| Gene Symbol: LOC105372582 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135006.1 | 1 | 25 | |||
| GenBank mRNA | DB215985.1 | - | 1 | 25 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000376445.3 | - | 1 | 15 | Chr. X - 48973723 - 49002264 [-] on Build GRCh38 | |

LOC105372582
Genomic Map