siRNA Details
| Gene Symbol: ZNRF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011530435.2 | XP_011528737.1 | 2 | 120 | ||
| XM_011530436.2 | XP_011528738.1 | 3 | 175 | |||
| XM_011530437.1 | XP_011528739.1 | 2 | 845 | |||
| XM_011530438.2 | XP_011528740.1 | 2 | 361 | |||
| XM_017028990.1 | XP_016884479.1 | 2 | 616 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000422972.1 | - | 2 | 204 | Chr. 22 - 28818331 - 28822215 [+] on Build GRCh38 | |

ZNRF3
Genomic Map