siRNA Details
| Gene Symbol: LOC101928627 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110412.1 | 1 | 269 | |||
| GenBank mRNA | AK092146.1 | - | 1 | 271 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000455438.2 | - | 1 | 269 | Chr. X - 36365626 - 36440292 [-] on Build GRCh38 | |

LOC101928627
Genomic Map