siRNA Details
| Gene Symbol: LINC00323 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024100.1 | 1 | 49 | |||
| GenBank mRNA | AK094492.1 | - | 1 | 49 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000441268.2 | - | 1 | 118 | Chr. 21 - 41141500 - 41148133 [-] on Build GRCh38 | |

LINC00323
Genomic Map