siRNA Details
| Gene Symbol: LOC101928280 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125854.1 | 4 | 437 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000445346.1 | - | 4 | 437 | Chr. 6 - 68226972 - 68329899 [-] on Build GRCh38 | |

LOC101928280
Genomic Map