siRNA Details
| Gene Symbol: LOC101928280 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125854.1 | 4 | 512 | |||
| GenBank mRNA | BC031312.2 | - | 4 | 514 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928280| Gene Symbol: LOC101928280 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125854.1 | 4 | 512 | |||
| GenBank mRNA | BC031312.2 | - | 4 | 514 | ||