siRNA Details
| Gene Symbol: LINC00343 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046391.2 | 2 | 549 | |||
| NR_120418.1 | 2 | 549 | ||||
| GenBank mRNA | AK058053.1 | - | 2 | 510 | ||
| BC126908.1 | - | 2 | 488 | |||
| BC127249.1 | - | 2 | 488 | |||
| CD244335.1 | - | 2 | 528 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000454555.2 | - | 2 | 734 | Chr. 13 - 105706645 - 105762100 [+] on Build GRCh38 | |

LINC00343
Genomic Map