siRNA Details
| Gene Symbol: LINC00974 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038442.1 | 3 | 1815 | |||
| GenBank mRNA | AK056318.1 | - | 3 | 1815 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000456403.1 | - | 3 | 1815 | Chr. 17 - 41549606 - 41554495 [-] on Build GRCh38 | |

LINC00974
Genomic Map