siRNA Details
Gene Symbol: LINC01108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108096.1 | 3 | 1094 | |||
NR_108097.1 | 2 | 858 | ||||
GenBank mRNA | AK056826.1 | - | 2 | 549 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000427276.1 | - | 2 | 549 | Chr. 6 - 14280127 - 14285145 [-] on Build GRCh38 |
