siRNA Details
| Gene Symbol: LINC01535 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110718.1 | 4 | 547 | |||
| NR_110720.1 | 3 | 427 | ||||
| GenBank mRNA | BC041478.1 | - | 4 | 506 | ||
| BX099288.1 | - | 3 | 389 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01535| Gene Symbol: LINC01535 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110718.1 | 4 | 547 | |||
| NR_110720.1 | 3 | 427 | ||||
| GenBank mRNA | BC041478.1 | - | 4 | 506 | ||
| BX099288.1 | - | 3 | 389 | |||