siRNA Details
| Gene Symbol: LINC01535 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110718.1 | 5 | 611 | |||
| GenBank mRNA | BC041478.1 | - | 5 | 570 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000592712.5 | - | 5 | 570 | Chr. 19 - 37251926 - 37263613 [+] on Build GRCh38 | |

LINC01535
Genomic Map