siRNA Details
| Gene Symbol: ENTPD1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134322.1 | 3 | 388 | |||
| GenBank mRNA | AL120918.1 | - | 3 | 326 | ||
| BU852745.1 | - | 3 | 391 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ENTPD1-AS1| Gene Symbol: ENTPD1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134322.1 | 3 | 388 | |||
| GenBank mRNA | AL120918.1 | - | 3 | 326 | ||
| BU852745.1 | - | 3 | 391 | |||