siRNA Details
| Gene Symbol: LOC101928525 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121579.1 | 3 | 837 | |||
| GenBank mRNA | BC015688.1 | - | 3 | 837 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000415062.1 | - | 3 | 837 | Chr. 9 - 135503273 - 135506447 [-] on Build GRCh38 | |

LOC101928525
Genomic Map