siRNA Details
| Gene Symbol: LOC100507657 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134581.1 | 2 | 4852 | |||
| GenBank mRNA | BX102824.1 | - | 1 | 162 | ||
| CR627208.1 | - | 2 | 4851 | |||
| CR936633.1 | - | 2 | 4851 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000426467.1 | - | 2 | 4852 | Chr. 22 - 27310651 - 27317456 [+] on Build GRCh38 | |

LOC100507657
Genomic Map