siRNA Details
| Gene Symbol: PSMD5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024408.1 | 4 | 1242 | |||
| GenBank mRNA | AL117474.1 | - | 3 | 966 | ||
| BC028086.1 | - | 4 | 1184 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PSMD5-AS1| Gene Symbol: PSMD5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024408.1 | 4 | 1242 | |||
| GenBank mRNA | AL117474.1 | - | 3 | 966 | ||
| BC028086.1 | - | 4 | 1184 | |||