siRNA Details
Gene Symbol: LOC100506682 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110024.1 | 4 | 524 | |||
GenBank mRNA | BX095908.1 | - | 2 | 341 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000448311.1 | - | 4 | 524 | Chr. 7 - 127476883 - 127485804 [+] on Build GRCh38 |
