siRNA Details
Gene Symbol: LOC339529 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033883.1 | 9 | 1111 | |||
GenBank mRNA | BC046644.1 | - | 9 | 1113 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000440494.1 | - | 9 | 1111 | Chr. 1 - 243917402 - 244047317 [+] on Build GRCh38 |
