siRNA Details
| Gene Symbol: LOC107984282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017017028.1 | XP_016872517.1 | 3 | 6861 | ||
| GenBank mRNA | AF086154.1 | - | 2 | 254 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000455414.1 | - | 2 | 248 | Chr. 10 - 132511626 - 132518191 [-] on Build GRCh38 | |

LOC107984282
Genomic Map