siRNA Details
Gene Symbol: LOC107984282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017017028.1 | XP_016872517.1 | 3 | 7145 | ||
GenBank mRNA | AF086154.1 | - | 2 | 538 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000455414.1 | - | 2 | 532 | Chr. 10 - 132511626 - 132518191 [-] on Build GRCh38 |
