siRNA Details
Gene Symbol: LINC00354 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046992.1 | 3 | 196 | |||
GenBank mRNA | BC032817.1 | - | 1 | 394 | ||
BX117816.1 | - | 3 | 188 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00354 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046992.1 | 3 | 196 | |||
GenBank mRNA | BC032817.1 | - | 1 | 394 | ||
BX117816.1 | - | 3 | 188 |