siRNA Details
| Gene Symbol: LINC00354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046992.1 | 3 | 315 | |||
| NR_120401.1 | 4 | 451 | ||||
| GenBank mRNA | BX117816.1 | - | 3 | 307 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000440860.1 | - | 2 | 248 | Chr. 13 - 111899985 - 111901176 [+] on Build GRCh38 | |

LINC00354
Genomic Map