siRNA Details
Gene Symbol: LINC00354 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046992.1 | 2 | 140 | |||
NR_120401.1 | 2 | 148 | ||||
GenBank mRNA | BQ186748.1 | - | 2 | 137 | ||
BX117816.1 | - | 2 | 132 | |||
HY022991.1 | - | 2 | 148 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000440860.1 | - | 1 | 73 | Chr. 13 - 111899985 - 111901176 [+] on Build GRCh38 |
