siRNA Details
Gene Symbol: LINC01276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY927638.1 | - | LIT3619 AY927638 |
1 | 139 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000448211.1 | - | 1 | 139 | Chr. 6 - 41518373 - 41519852 [+] on Build GRCh38 |
