siRNA Details
| Gene Symbol: LINC00161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026552.1 | 1 | 533 | |||
| GenBank mRNA | AY063458.1 | - | 1 | 533 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000412526.5 | - | 1 | 533 | Chr. 21 - 28539318 - 28540355 [+] on Build GRCh38 | |

LINC00161
Genomic Map