siRNA Details
| Gene Symbol: LINC01203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045260.1 | 4 | 187 | |||
| GenBank mRNA | AA470018.1 | - | 4 | 187 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000419566.5 | - | 4 | 187 | Chr. X - 13335241 - 13341825 [+] on Build GRCh38 | |

LINC01203
Genomic Map