siRNA Details
Gene Symbol: LINC01203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045260.1 | 6 | 473 | |||
GenBank mRNA | AA470018.1 | - | 6 | 473 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000419566.5 | - | 6 | 473 | Chr. X - 13335241 - 13341825 [+] on Build GRCh38 |
