siRNA Details
| Gene Symbol: LINC00658 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038239.1 | 5 | 1061 | |||
| NR_038240.1 | 5 | 1039 | ||||
| GenBank mRNA | BC035192.1 | - | 5 | 1066 | ||
| BC048109.1 | - | 5 | 1012 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000420529.5 | - | 5 | 999 | Chr. 20 - 5431992 - 5445686 [-] on Build GRCh38 | |

LINC00658
Genomic Map