siRNA Details
| Gene Symbol: SLC8A1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038441.1 | 4 | 161 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC8A1-AS1| Gene Symbol: SLC8A1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038441.1 | 4 | 161 | |||