siRNA Details
Gene Symbol: LINC00841 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136148.1 | 2 | 266 | |||
GenBank mRNA | AA432001.1 | - | 2 | 224 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000446107.1 | - | 2 | 224 | Chr. 10 - 43912444 - 43914236 [+] on Build GRCh38 |
