siRNA Details
Gene Symbol: FOCAD-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121601.1 | 2 | 335 | |||
GenBank mRNA | BX115825.1 | - | 2 | 335 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000439876.1 | - | 2 | 273 | Chr. 9 - 20683304 - 20684688 [-] on Build GRCh38 |
