siRNA Details
| Gene Symbol: LINC00595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073447.1 | 1 | 109 | |||
| GenBank mRNA | AK126491.1 | - | 1 | 95 | ||
| BX280027.1 | - | 1 | 33 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00595| Gene Symbol: LINC00595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073447.1 | 1 | 109 | |||
| GenBank mRNA | AK126491.1 | - | 1 | 95 | ||
| BX280027.1 | - | 1 | 33 | |||