siRNA Details
| Gene Symbol: WDR11-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC043378.1 | - | 3 | 427 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000451706.3 | - | 4 | 592 | Chr. 10 - 120763401 - 120793360 [-] on Build GRCh38 | |

WDR11-AS1
Genomic Map