siRNA Details
Gene Symbol: WDR11-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC043378.1 | - | 2, 3 | 231 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000451706.3 | - | 3 | 396 | Chr. 10 - 120763401 - 120793360 [-] on Build GRCh38 |
