siRNA Details
| Gene Symbol: MGAT3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126469.1 | 1 | 335 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000432239.1 | - | 1 | 335 | Chr. 22 - 39475807 - 39476822 [-] on Build GRCh38 | |

MGAT3-AS1
Genomic Map