siRNA Details
| Gene Symbol: LOC101928222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110235.1 | 3 | 497 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000416575.1 | - | 3 | 484 | Chr. 2 - 19990217 - 20004210 [+] on Build GRCh38 | |

LOC101928222
Genomic Map