siRNA Details
Gene Symbol: LINC00845 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108052.1 | 2 | 607 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000417931.1 | - | 2 | 607 | Chr. 10 - 61016275 - 61026420 [+] on Build GRCh38 |
