siRNA Details
Gene Symbol: SMIM2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104064.1 | 1 | 235 | |||
GenBank mRNA | DB036693.1 | - | 1 | 235 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000444663.5 | - | 1 | 17 | Chr. 13 - 44142546 - 44148030 [+] on Build GRCh38 |
