siRNA Details
Gene Symbol: LINC01268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038863.2 | 3 | 2081 | |||
GenBank mRNA | BC037331.1 | - | 3 | 2086 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000434296.2 | - | 3 | 2085 | Chr. 6 - 113868013 - 113873351 [-] on Build GRCh38 |
