siRNA Details
| Gene Symbol: LINC01624 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104177.1 | 2 | 2978 | |||
| GenBank mRNA | BX088681.1 | - | 2 | 139 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000438622.1 | - | 2 | 106 | Chr. 6 - 170276939 - 170279469 [+] on Build GRCh38 | |

LINC01624
Genomic Map