siRNA Details
| Gene Symbol: LINC01624 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104177.1 | 1 | 1868 | |||
| GenBank mRNA | AK095441.1 | - | 1 | 1868 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000607074.1 | - | 1 | 1868 | Chr. 6 - 170272474 - 170279887 [+] on Build GRCh38 | |

LINC01624
Genomic Map