siRNA Details
| Gene Symbol: LINC01483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC068583.1 | - | 4 | 2184 | ||
| BC094791.1 | - | 4 | 2184 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000433856.5 | - | 3 | 2166 | Chr. 17 - 69594785 - 69628351 [+] on Build GRCh38 | |

LINC01483
Genomic Map