siRNA Details
Gene Symbol: LINC01483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC068583.1 | - | 4 | 1239 | ||
BC094791.1 | - | 4 | 1239 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000433856.5 | - | 3 | 1221 | Chr. 17 - 69594785 - 69628351 [+] on Build GRCh38 |
