siRNA Details
Gene Symbol: LOC100506725 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108082.1 | 3 | 651 | |||
NR_108083.1 | 2 | 553 | ||||
GenBank mRNA | HY127409.1 | - | 2 | 263 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100506725 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108082.1 | 3 | 651 | |||
NR_108083.1 | 2 | 553 | ||||
GenBank mRNA | HY127409.1 | - | 2 | 263 |