siRNA Details
Gene Symbol: LINC00355 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057471.1 | - | 6 | 1648 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000456627.1 | - | 6 | 1648 | Chr. 13 - 63986371 - 64076011 [-] on Build GRCh38 |
