siRNA Details
| Gene Symbol: LOC105748977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131925.1 | 2 | 201 | |||
| GenBank mRNA | BF726453.1 | - | 2 | 201 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105748977| Gene Symbol: LOC105748977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131925.1 | 2 | 201 | |||
| GenBank mRNA | BF726453.1 | - | 2 | 201 | ||